siRNA Details
Gene Symbol: TDRD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153046.2 | NP_694591.2 | 7 | 932 | ||
XM_005267309.3 | XP_005267366.1 | 7 | 1069 | |||
XM_006720019.3 | XP_006720082.1 | 7 | 1069 | |||
XM_006720020.3 | XP_006720083.1 | 6 | 988 | |||
XM_011536397.2 | XP_011534699.1 | 7 | 1069 | |||
XM_011536398.2 | XP_011534700.1 | 7 | 874 | |||
XM_011536400.2 | XP_011534702.1 | 7 | 1069 | |||
XM_011536401.2 | XP_011534703.1 | 7 | 807 | |||
XM_017020955.1 | XP_016876444.1 | 7 | 923 | |||
GenBank mRNA | AK125978.1 | - | 2 | 81 | ||
AL833915.1 | - | 4 | 322 | |||
BC128057.1 | - | 2 | 71 | 40076927 | ||
U56244.1 | - | 6 | 671 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
