siRNA Details
Gene Symbol: ZNF761 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008401.3 | NP_001008401.3 | 6 | 850 | ||
NM_001289951.1 | NP_001276880.1 | 5 | 631 | |||
NM_001289952.1 | NP_001276881.1 | 5 | 699 | |||
GenBank mRNA | AB107355.2 | - | 5 | 700 | ||
AK131281.1 | - | 2 | 320 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
