siRNA Details
Gene Symbol: ZNF761 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008401.3 | NP_001008401.3 | 6 | 949 | ||
NM_001289951.1 | NP_001276880.1 | 5 | 730 | |||
NM_001289952.1 | NP_001276881.1 | 5 | 798 | |||
GenBank mRNA | AB107355.2 | - | 5 | 799 | ||
AK131281.1 | - | 2 | 419 |
Gene Symbol: ZNF765 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040185.1 | NP_001035275.1 | 4 | 614 | ||
GenBank mRNA | AK293890.1 | - | 6 | 828 | ||
AK316300.1 | - | 4 | 608 | |||
BC140724.1 | - | 4 | 588 | 9021592 | ||
BX647981.1 | - | 4 | 606 | |||
CD656338.1 | - | 4 | 635 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
