siRNA Details
Gene Symbol: TCP10L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144659.5 | NP_653260.1 | 5 | 1963 | ||
GenBank mRNA | AK001794.1 | - | 1 | 307 | ||
BC027970.1 | - | 1 | 717 | 5225527 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
