siRNA Details
Gene Symbol: MEGF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271938.1 | NP_001258867.1 | 42 | 11070 | ||
NM_001410.2 | NP_001401.2 | 41 | 10869 | |||
GenBank mRNA | AF497566.1 | - | 1 | 280 | ||
AK026018.1 | - | 1 | 696 | |||
BC093649.1 | - | 1 | 306 | |||
BC111942.1 | - | 1 | 306 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
