siRNA Details
Gene Symbol: KGFLP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003670.1 | 3 | 1590 | |||
GenBank mRNA | AY098594.1 | - | 3 | 1482 |
Gene Symbol: KGFLP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003674.2 | 3 | 1491 | |||
NR_047527.1 | 4 | 1601 | ||||
GenBank mRNA | AF523265.1 | - | 3 | 1586 | ||
AY098593.1 | - | 3 | 1482 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
