siRNA Details
Gene Symbol: KGFLP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003674.2 | 1 | 309 | |||
NR_047527.1 | 1 | 309 | ||||
GenBank mRNA | AF523265.1 | - | 1 | 404 | ||
AK054997.1 | - | 1 | 48 | |||
AK307215.1 | - | 1 | 40 | |||
AY098593.1 | - | 1 | 300 | |||
BC101842.1 | - | 1 | 309 | |||
BC112977.1 | - | 1 | 307 | |||
BC112978.1 | - | 1 | 307 |
Gene Symbol: KGFLP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003670.1 | 1 | 408 |
Gene Symbol: LOC102724238 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK290223.1 | - | 1 | 316 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
