siRNA Details
Gene Symbol: HLA-DQB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243961.1 | NP_001230890.1 | 1, 2 | 190 | ||
NM_001243962.1 | NP_001230891.1 | 1, 2 | 187 | |||
NM_002123.4 | NP_002114.3 | 1, 2 | 190 | |||
GenBank mRNA | AB209580.1 | - | 1, 2 | 139 | ||
AB209638.1 | - | 1, 2 | 167 | |||
AF195245.1 | - | 12 | ||||
AF291674.1 | - | 1, 2 | 108 | |||
AK295255.1 | - | 1, 2 | 155 | |||
AK295476.1 | - | 1, 2 | 160 | |||
AK295695.1 | - | 1, 2 | 158 | |||
AK296060.1 | - | 1, 2 | 152 | |||
AK296613.1 | - | 1, 2 | 169 | |||
AK304543.1 | - | 1, 2 | 194 | |||
AK316333.1 | - | 1, 2 | 182 | |||
AY656681.1 | - | 1, 2 | 108 | |||
AY656682.1 | - | 1, 2 | 108 | |||
AY656683.1 | - | 1, 2 | 108 | |||
BC012106.1 | - | 1, 2 | 162 | 4574913 | ||
DC409119.1 | - | 1, 2 | 190 | |||
DQ136036.1 | - | 1, 2 | 153 | |||
J00198.1 | - | 17 | ||||
L34095.1 | - | 12 | ||||
L34096.1 | - | 12 | ||||
L34097.1 | - | 12 | ||||
L34098.1 | - | 12 | ||||
L34099.1 | - | 12 | ||||
L34100.1 | - | 12 | ||||
L34101.1 | - | 12 | ||||
L34102.1 | - | 12 | ||||
L34103.1 | - | 12 | ||||
L34105.1 | - | 12 | ||||
L34106.1 | - | 12 | ||||
L34107.1 | - | 12 | ||||
L40179.1 | - | 12 | ||||
L40180.1 | - | 12 | ||||
L42626.1 | - | 12 | ||||
M13279.1 | - | 12 | ||||
M14189.1 | - | 12 | ||||
M16996.1 | - | 1, 2 | 108 | |||
M17206.1 | - | 12 | ||||
M17207.1 | - | 12 | ||||
M17563.1 | - | 1, 2 | 120 | |||
M17564.1 | - | 1, 2 | 120 | |||
M17565.1 | - | 1, 2 | 76 | |||
M17955.1 | - | 1, 2 | 180 | |||
M19239.1 | - | 12 | ||||
M20432.1 | - | 1, 2 | 167 | |||
M20505.1 | - | 1, 2 | 76 | |||
M21135.1 | - | 12 | ||||
M22549.1 | - | 12 | ||||
M22550.1 | - | 12 | ||||
M24364.1 | - | 1, 2 | 152 | |||
M25325.1 | - | 1, 2 | 190 | |||
M25326.1 | - | 1, 2 | 150 | |||
M25327.1 | - | 1, 2 | 185 | |||
M26042.1 | - | 1, 2 | 153 | |||
M32577.1 | - | 1, 2 | 76 | |||
M33907.1 | - | 1, 2 | 153 | |||
M35982.1 | - | 1, 2 | 91 | |||
M59800.1 | - | 12 | ||||
M59801.1 | - | 12 | ||||
M60028.1 | - | 1, 2 | 165 | |||
M81140.1 | - | 1, 2 | 129 | |||
M81141.1 | - | 1, 2 | 143 | |||
M83770.1 | - | 12 | ||||
U66400.1 | - | 12 | ||||
U83582.1 | - | 1, 2 | 106 | |||
U83585.1 | - | 1, 2 | 106 | |||
V00529.1 | - | 17 | ||||
X00369.1 | - | 1, 2 | 76 | |||
X03068.1 | - | 1, 2 | 189 | |||
X76553.1 | - | 12 | ||||
X76554.1 | - | 12 | ||||
X96420.1 | - | 1, 2 | 108 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
