siRNA Details
Gene Symbol: HLA-DQB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243961.1 | NP_001230890.1 | 2 | 408 | ||
NM_002123.4 | NP_002114.3 | 2 | 408 | |||
GenBank mRNA | AB209580.1 | - | 2 | 357 | ||
AB209638.1 | - | 2 | 385 | |||
AF000447.1 | - | 2 | 319 | |||
AF113250.1 | - | 1 | 224 | |||
AF195245.1 | - | 1 | 230 | |||
AK295476.1 | - | 2 | 378 | |||
AK295695.1 | - | 2 | 376 | |||
AK296613.1 | - | 2 | 387 | |||
AK316333.1 | - | 2 | 400 | |||
AY656682.1 | - | 2 | 326 | |||
BC012106.1 | - | 2 | 380 | 4574913 | ||
BC128595.1 | - | 1 | 222 | |||
DC409119.1 | - | 2 | 408 | |||
DQ136036.1 | - | 2 | 371 | |||
KC879155.1 | - | 1 | 218 | |||
L34096.1 | - | 1 | 230 | |||
L34097.1 | - | 1 | 230 | |||
L34098.1 | - | 1 | 230 | |||
L34104.1 | - | 1 | 230 | |||
L34105.1 | - | 1 | 230 | |||
L34106.1 | - | 1 | 230 | |||
L34107.1 | - | 1 | 230 | |||
L40181.1 | - | 1 | 230 | |||
L42626.1 | - | 1 | 230 | |||
M16276.1 | - | 2 | 536 | |||
M16996.1 | - | 2 | 326 | |||
M17204.1 | - | 1 | 230 | |||
M17207.1 | - | 1 | 230 | |||
M17563.1 | - | 2 | 338 | |||
M17955.1 | - | 2 | 398 | |||
M20432.1 | - | 2 | 385 | |||
M20505.1 | - | 2 | 294 | |||
M21135.1 | - | 1 | 230 | |||
M22550.1 | - | 1 | 230 | |||
M24364.1 | - | 2 | 370 | |||
M25325.1 | - | 2 | 408 | |||
M25326.1 | - | 2 | 368 | |||
M25327.1 | - | 2 | 403 | |||
M25328.1 | - | 1 | 136 | |||
M32577.1 | - | 2 | 294 | |||
M35730.1 | - | 1 | 217 | |||
M35731.1 | - | 1 | 217 | |||
M57649.1 | - | 2 | 312 | |||
M57650.1 | - | 1 | 72 | |||
M59800.1 | - | 1 | 230 | |||
M59801.1 | - | 1 | 230 | |||
M60028.1 | - | 2 | 383 | |||
M83770.1 | - | 1 | 230 | |||
U63321.1 | - | 1 | 224 | |||
U66400.1 | - | 1 | 230 | |||
U83582.1 | - | 2 | 324 | |||
U83585.1 | - | 2 | 324 | |||
X76553.1 | - | 1 | 230 | |||
X76554.1 | - | 1 | 230 | |||
X96420.1 | - | 2 | 326 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
