siRNA Details
Gene Symbol: HLA-DQB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243961.1 | NP_001230890.1 | 4 | 839 | ||
NM_002123.4 | NP_002114.3 | 4 | 839 | |||
GenBank mRNA | AB209580.1 | - | 2 | 3846 | ||
AF000447.1 | - | 4 | 750 | |||
AK296613.1 | - | 4 | 818 | |||
AK316333.1 | - | 4 | 831 | |||
BC128595.1 | - | 3 | 653 | |||
L34104.1 | - | 3 | 661 | |||
L34105.1 | - | 3 | 661 | |||
L34106.1 | - | 3 | 661 | |||
L34107.1 | - | 3 | 661 | |||
L40181.1 | - | 3 | 661 | |||
L42626.1 | - | 3 | 661 | |||
M16276.1 | - | 4 | 967 | |||
M17563.1 | - | 4 | 769 | |||
M20432.1 | - | 4 | 816 | |||
M57649.1 | - | 4 | 743 | |||
M57650.1 | - | 3 | 503 | |||
X96420.1 | - | 4 | 757 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
