siRNA Details
Gene Symbol: C14orf80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134875.1 | NP_001128347.1 | 6, 7 | 909 | ||
NM_001134876.1 | NP_001128348.1 | 5, 6 | 810 | |||
NM_001134877.1 | NP_001128349.1 | 6, 7 | 737 | |||
NM_001198983.1 | NP_001185912.1 | 6, 7 | 819 | |||
GenBank mRNA | AK024506.1 | - | 1 | 3651 | ||
BC016028.1 | - | 5, 6 | 577 | 3907551 | ||
BX161477.1 | - | 6, 7 | 819 | |||
BX248038.1 | - | 6, 7 | 737 | |||
BX248074.1 | - | 7, 8 | 1037 | |||
BX248283.1 | - | 5, 6 | 768 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
