siRNA Details
Gene Symbol: RPS19BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194326.3 | NP_919307.1 | 2, 3 | 308 | ||
NR_130151.1 | 3, 4 | 399 | ||||
GenBank mRNA | AK025423.1 | - | 2, 3 | 201 | ||
AK055865.1 | - | 1 | 2186 | |||
BC037573.1 | - | 2, 3 | 318 | 5399759 | ||
BC047711.1 | - | 2, 3 | 200 | 6064917 | ||
CB142920.1 | - | 3, 4 | 305 | |||
CR456443.1 | - | 2, 3 | 238 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
