siRNA Details
Gene Symbol: OR13F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004485.1 | NP_001004485.1 | 1 | 412 | ||
GenBank mRNA | BC140739.1 | - | 1 | 501 | 9021607 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
