siRNA Details
Gene Symbol: C9orf85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182505.3 | NP_872311.2 | 3 | 476 | ||
XM_011518226.2 | XP_011516528.1 | 4 | 774 | |||
GenBank mRNA | BC010179.2 | - | 2 | 365 | ||
BC052375.1 | - | 3 | 315 | 6147448 | ||
BQ431408.1 | - | 3 | 277 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
