siRNA Details
Gene Symbol: C9orf85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182505.3 | NP_872311.2 | 1 | 252 | ||
XM_011518226.2 | XP_011516528.1 | 1 | 419 | |||
GenBank mRNA | AK307186.1 | - | 1 | 65 | ||
AK311206.1 | - | 1 | 120 | |||
BC010179.2 | - | 1 | 248 | |||
BC028019.1 | - | 1 | 98 | |||
BC052375.1 | - | 1 | 91 | 6147448 | ||
BQ431408.1 | - | 1 | 53 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
