siRNA Details
Gene Symbol: KIR3DX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026716.2 | 9 | 1323 | |||
NR_104095.1 | 8 | 1003 | ||||
NR_104096.1 | 8 | 985 | ||||
NR_104097.1 | 8 | 1096 | ||||
NR_136268.1 | 9 | 1323 | ||||
GenBank mRNA | AK024467.1 | - | 4 | 2143 | ||
BC033195.1 | - | 4 | 358 | |||
BC171827.1 | - | 8 | 1003 | |||
BC171832.1 | - | 8 | 985 | |||
BC171834.1 | - | 8 | 1257 | |||
BC171838.1 | - | 8 | 1096 | |||
BC171839.1 | - | 7 | 1198 | |||
DQ224421.1 | - | 9 | 1224 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
