siRNA Details
Gene Symbol: LOC647264 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC157826.1 | - | 460 | 9057045 | ||
BC171812.1 | - | 460 | 9054466 |
Gene Symbol: KRTAP21-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181619.1 | NP_853650.1 | 1 | 226 | ||
GenBank mRNA | AB096960.1 | - | 1 | 255 | ||
BC105029.1 | - | 1 | 276 | 8144032 | ||
BC105031.1 | - | 1 | 276 | 8144034 |
Gene Symbol: LINC01198 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC046635.1 | - | 1 | 133 |
Gene Symbol: LOC101927016 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011535339.1 | XP_011533641.1 | 1 | 1043 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
