siRNA Details
Gene Symbol: DCDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195610.1 | NP_001182539.1 | 2 | 337 | ||
NM_016356.4 | NP_057440.2 | 1 | 430 | |||
GenBank mRNA | AB032980.2 | - | 1 | 128 | ||
AF181721.1 | - | 1 | 430 | |||
AK027036.1 | - | 1 | 246 | |||
BC050704.1 | - | 2 | 337 | 5758845 | ||
BM923267.1 | - | 2 | 350 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
