siRNA Details
Gene Symbol: C19orf25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152482.2 | NP_689695.2 | 3 | 461 | ||
XM_005259506.3 | XP_005259563.1 | 3 | 477 | |||
XM_006722653.3 | XP_006722716.1 | 3 | 436 | |||
XM_017026374.1 | XP_016881863.1 | 3 | 784 | |||
GenBank mRNA | AK056004.1 | - | 1 | 1489 | ||
AK075267.1 | - | 3 | 444 | |||
AK093985.1 | - | 4 | 338 | |||
AK297944.1 | - | 3 | 568 | |||
AL122089.1 | - | 3 | 432 | |||
AY007120.1 | - | 2 | 397 | |||
BC018441.1 | - | 3 | 431 | 4471390 | ||
BP210473.1 | - | 3 | 461 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
