siRNA Details
Gene Symbol: CYP27C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001665.3 | NP_001001665.3 | 7 | 1040 | ||
XM_017003960.1 | XP_016859449.1 | 8 | 1230 | |||
XM_017003961.1 | XP_016859450.1 | 7 | 1118 | |||
GenBank mRNA | AK131190.1 | - | 7 | 1021 | ||
BC039307.1 | - | 7 | 1040 | 5266370 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
