siRNA Details
Gene Symbol: CYP27C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001665.3 | NP_001001665.3 | 2, 3 | 303 | ||
XM_017003960.1 | XP_016859449.1 | 3, 4 | 493 | |||
XM_017003961.1 | XP_016859450.1 | 1, 2 | 194 | |||
GenBank mRNA | AK131190.1 | - | 2, 3 | 284 | ||
BC039307.1 | - | 2, 3 | 303 | 5266370 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
