siRNA Details
Gene Symbol: DLEU7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306135.1 | NP_001293064.1 | 1 | 730 | ||
NM_198989.3 | NP_945340.2 | 1 | 730 | |||
GenBank mRNA | AK126830.1 | - | 1 | 730 | ||
AK303567.1 | - | 2 | 547 | |||
AY357595.1 | - | 1 | 487 | |||
BC104892.1 | - | 1 | 540 | 8143895 | ||
BC112009.1 | - | 1 | 540 | 8327477 | ||
DN992279.1 | - | 1 | 313 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
