siRNA Details
Gene Symbol: CFAP206 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031743.2 | NP_001026913.1 | 2 | 221 | ||
GenBank mRNA | AK093830.1 | - | 2 | 180 | ||
BC035083.2 | - | 2 | 190 | 5260875 | ||
DB021694.1 | - | 2 | 217 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
