siRNA Details
Gene Symbol: LCE3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178431.1 | NP_848518.1 | 1 | 182 | ||
GenBank mRNA | BC120933.1 | - | 1 | 182 | ||
BC120934.1 | - | 1 | 182 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
