siRNA Details
Gene Symbol: THAP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130475.2 | NP_001123947.1 | 3 | 1417 | ||
NM_001287598.1 | NP_001274527.1 | 3 | 1371 | |||
NM_001287599.1 | NP_001274528.1 | 3 | 1295 | |||
NM_001287601.1 | NP_001274530.1 | 2 | 1224 | |||
NM_182529.3 | NP_872335.2 | 3 | 1493 | |||
GenBank mRNA | AL833137.1 | - | 3 | 1372 | ||
BC053634.1 | - | 3 | 1493 | 5165075 |
Gene Symbol: THAP5P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126609.1 | - | 1 | 2694 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
