siRNA Details
Gene Symbol: IL18BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039659.1 | NP_001034748.1 | 4 | 321 | ||
NM_001039660.1 | NP_001034749.1 | 3 | 209 | |||
NM_001145055.1 | NP_001138527.1 | 2 | 101 | |||
NM_001145057.1 | NP_001138529.1 | 3 | 147 | |||
NM_005699.3 | NP_005690.2 | 2 | 965 | |||
NM_173042.2 | NP_766630.2 | 2 | 965 | |||
NM_173044.2 | NP_766632.2 | 3 | 209 | |||
XM_017017059.1 | XP_016872548.1 | 3 | 911 | |||
XM_017017060.1 | XP_016872549.1 | 3 | 878 | |||
XM_017017061.1 | XP_016872550.1 | 4 | 774 | |||
XM_017017062.1 | XP_016872551.1 | 4 | 647 | |||
XM_017017063.1 | XP_016872552.1 | 4 | 1223 | |||
GenBank mRNA | AB019504.1 | - | 2 | 43 | ||
AF110799.1 | - | 2 | 102 | |||
AF110800.1 | - | 2 | 102 | |||
AF110801.1 | - | 2 | 965 | |||
AF122906.1 | - | 3 | 121 | |||
AF215907.2 | - | 3 | 198 | |||
AK098255.1 | - | 2 | 2059 | |||
BC044215.1 | - | 4 | 303 | 5755934 | ||
BX357194.2 | - | 4 | 341 | |||
CB154498.1 | - | 3 | 212 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
