siRNA Details
Gene Symbol: FNDC3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079673.1 | NP_001073141.1 | 13, 14 | 1839 | ||
NM_001278438.1 | NP_001265367.1 | 13, 14 | 1882 | |||
NM_014923.4 | NP_055738.3 | 11, 12 | 1463 | |||
NR_103528.1 | 13, 14 | 1882 | ||||
XM_011534997.2 | XP_011533299.1 | 12, 13 | 1553 | |||
XM_017020440.1 | XP_016875929.1 | 13, 14 | 1724 | |||
XM_017020441.1 | XP_016875930.1 | 15, 16 | 2345 | |||
XM_017020442.1 | XP_016875931.1 | 14, 15 | 1877 | |||
XM_017020443.1 | XP_016875932.1 | 13, 14 | 2197 | |||
GenBank mRNA | AB023187.2 | - | 11, 12 | 1393 | ||
AJ749706.1 | - | 13, 14 | 1841 | |||
AJ749707.1 | - | 11, 12 | 1454 | |||
AK302415.1 | - | 11, 12 | 1463 | |||
BC060816.1 | - | 4, 5 | 501 | |||
BC070072.1 | - | 4, 5 | 501 | |||
BC107793.1 | - | 10, 11 | 1330 | |||
BC132812.1 | - | 13, 14 | 1606 | 40146834 | ||
BC136617.1 | - | 13, 14 | 1607 | 9020607 | ||
BC144301.1 | - | 13, 14 | 1607 | 9052821 | ||
BX640739.1 | - | 13, 14 | 1742 | |||
BX648141.1 | - | 13, 14 | 1829 | |||
BX648706.1 | - | 15, 16 | 1687 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
