siRNA Details
Gene Symbol: FNDC3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079673.1 | NP_001073141.1 | 15 | 1958 | ||
NM_001278438.1 | NP_001265367.1 | 15 | 2001 | |||
NM_014923.4 | NP_055738.3 | 13 | 1582 | |||
NR_103528.1 | 15 | 2001 | ||||
XM_011534997.2 | XP_011533299.1 | 14 | 1672 | |||
XM_017020440.1 | XP_016875929.1 | 15 | 1843 | |||
XM_017020441.1 | XP_016875930.1 | 17 | 2464 | |||
XM_017020442.1 | XP_016875931.1 | 16 | 1996 | |||
XM_017020443.1 | XP_016875932.1 | 15 | 2316 | |||
GenBank mRNA | AB023187.2 | - | 13 | 1512 | ||
AJ749706.1 | - | 15 | 1960 | |||
AJ749707.1 | - | 13 | 1573 | |||
AK302415.1 | - | 13 | 1582 | |||
BC060816.1 | - | 6 | 620 | |||
BC070072.1 | - | 6 | 620 | |||
BC107793.1 | - | 12 | 1449 | |||
BC132812.1 | - | 15 | 1725 | 40146834 | ||
BC136617.1 | - | 15 | 1726 | 9020607 | ||
BC144301.1 | - | 15 | 1726 | 9052821 | ||
BX640739.1 | - | 15 | 1861 | |||
BX648141.1 | - | 15 | 1948 | |||
BX648706.1 | - | 17 | 1806 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
