siRNA Details
Gene Symbol: GJD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152219.3 | NP_689343.3 | 1 | 1075 | ||
GenBank mRNA | AK125254.1 | - | 1 | 1075 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
