siRNA Details
Gene Symbol: FAM101A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181709.4 | NP_859060.3 | 3 | 580 | ||
GenBank mRNA | AK074545.1 | - | 5, 6 | 493 | ||
AK126577.1 | - | 3 | 580 | |||
BC042102.1 | - | 3 | 487 | |||
BC141805.1 | - | 2 | 348 | 40147249 |
Gene Symbol: ZNF664-FAM101A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204299.1 | NP_001191228.1 | 5 | 843 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
