siRNA Details
Gene Symbol: OR2A14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001659.1 | NP_001001659.1 | 1 | 659 | ||
GenBank mRNA | BC136889.1 | - | 1 | 711 | 9020880 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
