siRNA Details
Gene Symbol: NBPF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101663.4 | NP_001095133.3 | 22 | 4232 | ||
NM_183372.5 | NP_899228.4 | 24 | 4926 | |||
NR_046188.3 | 25 | 4994 | ||||
NR_110864.1 | 23 | 4918 | ||||
GenBank mRNA | AK054850.1 | - | 8 | 2675 | ||
BX648497.1 | - | 23 | 4717 | |||
CN421512.1 | - | 1 | 587 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
