siRNA Details
Gene Symbol: IER5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203434.2 | NP_982258.2 | 1 | 682 | ||
GenBank mRNA | BC064028.1 | - | 1 | 668 | 6526871 | |
BX458317.2 | - | 1 | 649 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
