siRNA Details
Gene Symbol: PAGE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207339.3 | NP_997222.1 | 4 | 335 | ||
XM_017029353.1 | XP_016884842.1 | 4 | 364 | |||
GenBank mRNA | BX113994.1 | - | 4 | 265 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
