siRNA Details
Gene Symbol: PAGE2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015038.2 | NP_001015038.1 | 4, 5 | 390 | ||
XM_011530785.2 | XP_011529087.1 | 5, 6 | 806 | |||
XM_011530786.2 | XP_011529088.1 | 4, 5 | 473 | |||
XM_011530787.2 | XP_011529089.1 | 4, 5 | 453 | |||
XM_017029513.1 | XP_016885002.1 | 4, 5 | 410 | |||
GenBank mRNA | BC130359.1 | - | 4, 5 | 349 | 40146390 | |
BM782405.1 | - | 4, 5 | 375 |
Gene Symbol: PAGE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207339.3 | NP_997222.1 | 4, 5 | 383 | ||
XM_017029353.1 | XP_016884842.1 | 4, 5 | 412 | |||
GenBank mRNA | BC054022.1 | - | 4, 5 | 377 | 6603202 | |
BX113994.1 | - | 4, 5 | 313 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
