siRNA Details
Gene Symbol: HMX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306142.1 | NP_001293071.1 | 1 | 220 | ||
NM_018942.2 | NP_061815.2 | 1 | 220 | |||
GenBank mRNA | AB593128.1 | - | 1 | 131 | ||
M99587.1 | - | 1 | 216 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
