siRNA Details
Gene Symbol: HMX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018942.2 | NP_061815.2 | 1, 2 | 597 | ||
GenBank mRNA | M99587.1 | - | 1, 2 | 587 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
