siRNA Details
Gene Symbol: SYNE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039876.2 | NP_001034965.1 | 8 | 1307 | ||
NM_001297735.2 | NP_001284664.1 | 6 | 968 | |||
GenBank mRNA | AK093764.1 | - | 7 | 2266 | ||
BC052573.1 | - | 6 | 958 | 6568945 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
