siRNA Details
Gene Symbol: NAPRT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286829.1 | NP_001273758.1 | 10, 11 | 1331 | ||
NM_145201.5 | NP_660202.3 | 10, 11 | 1331 | |||
XM_017013975.1 | XP_016869464.1 | 9, 10 | 2893 | |||
XM_017013976.1 | XP_016869465.1 | 9, 10 | 2887 | |||
XM_017013977.1 | XP_016869466.1 | 9, 10 | 2593 | |||
XM_017013979.1 | XP_016869468.1 | 9, 10 | 1089 | |||
XM_017013980.1 | XP_016869469.1 | 9, 10 | 1194 | |||
GenBank mRNA | AB242230.1 | - | 10, 11 | 1318 | ||
AF258565.1 | - | 4, 5 | 779 | |||
AK097002.1 | - | 10, 11 | 1323 | |||
AY214325.1 | - | 10, 11 | 1319 | |||
AY214326.1 | - | 10, 11 | 1319 | |||
AY214327.1 | - | 10, 11 | 1319 | |||
BC006284.2 | - | 10, 11 | 1303 | 3957135 | ||
BC014190.2 | - | 10, 11 | 1303 | |||
BC032466.1 | - | 5, 6 | 617 | 5210589 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
