siRNA Details
Gene Symbol: NAPRT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286829.1 | NP_001273758.1 | 3 | 437 | ||
NM_145201.5 | NP_660202.3 | 3 | 437 | |||
XM_017013975.1 | XP_016869464.1 | 3 | 1780 | |||
XM_017013976.1 | XP_016869465.1 | 3 | 1774 | |||
XM_017013977.1 | XP_016869466.1 | 3 | 1780 | |||
XM_017013978.1 | XP_016869467.1 | 3 | 1780 | |||
XM_017013979.1 | XP_016869468.1 | 3 | 311 | |||
XM_017013980.1 | XP_016869469.1 | 3 | 416 | |||
GenBank mRNA | AB242230.1 | - | 3 | 424 | ||
AK097002.1 | - | 3 | 429 | |||
AY214325.1 | - | 3 | 425 | |||
AY214326.1 | - | 3 | 425 | |||
AY214327.1 | - | 3 | 425 | |||
BC006284.2 | - | 3 | 409 | 3957135 | ||
BC014190.2 | - | 3 | 409 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
