siRNA Details
Gene Symbol: UTS2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198152.3 | NP_937795.2 | 6 | 927 | ||
XM_011512631.1 | XP_011510933.1 | 5 | 592 | |||
XM_017006091.1 | XP_016861580.1 | 5 | 592 | |||
GenBank mRNA | AB116021.1 | - | 2 | 189 | ||
AK090630.1 | - | 6 | 926 | |||
AY321313.1 | - | 2 | 202 | |||
BC112166.1 | - | 2 | 185 | 8327634 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
