siRNA Details
Gene Symbol: HNF1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000545.6 | NP_000536.5 | 2 | 559 | ||
NM_001306179.1 | NP_001293108.1 | 2 | 559 | |||
XM_005253931.3 | XP_005253988.1 | 2 | 612 | |||
GenBank mRNA | AK298422.1 | - | 2 | 433 | ||
BC104908.1 | - | 2 | 378 | 8143911 | ||
BC104910.1 | - | 2 | 378 | 8143913 | ||
BC143483.1 | - | 2 | 378 | |||
HM116541.1 | - | 2 | 501 | |||
HM116542.1 | - | 2 | 501 | |||
HM116543.1 | - | 2 | 501 | |||
HM116544.1 | - | 2 | 501 | |||
HM116547.1 | - | 2 | 501 | |||
HM116548.1 | - | 2 | 501 | |||
HM116552.1 | - | 3 | 118 | |||
HM116557.1 | - | 2 | 476 | |||
HM116558.1 | - | 2 | 476 | |||
HM449088.1 | - | 2 | 381 | |||
M57732.1 | - | 2 | 381 | |||
X71346.1 | - | 2 | 554 | |||
X71347.1 | - | 2 | 489 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
