siRNA Details
Gene Symbol: HNF1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000545.6 | NP_000536.5 | 10 | 2036 | ||
NM_001306179.1 | NP_001293108.1 | 10 | 2057 | |||
XM_005253931.3 | XP_005253988.1 | 9 | 2182 | |||
GenBank mRNA | BC104908.1 | - | 10 | 1855 | 8143911 | |
BC104910.1 | - | 10 | 1855 | 8143913 | ||
BC143483.1 | - | 10 | 1877 | |||
HM116542.1 | - | 10 | 1652 | |||
HM116543.1 | - | 8 | 1441 | |||
HM116544.1 | - | 7 | 1095 | |||
HM116545.1 | - | 7 | 965 | |||
HM116546.1 | - | 5 | 595 | |||
HM116549.1 | - | 4 | 295 | |||
HM116551.1 | - | 2 | 151 | |||
HM449088.1 | - | 9 | 1951 | |||
HM449089.1 | - | 9 | 1658 | |||
M57732.1 | - | 10 | 1858 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
