siRNA Details
Gene Symbol: SPATA41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028139.1 | 1 | 723 | |||
NR_028140.1 | 1 | 238 | ||||
GenBank mRNA | AK124283.1 | - | 1 | 718 | ||
BC028144.1 | - | 1 | 234 | 5214282 | ||
BC132704.1 | - | 1 | 181 | |||
BC132706.1 | - | 1 | 181 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
