siRNA Details
Gene Symbol: SPATA41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028139.1 | 1 | 1168 | |||
GenBank mRNA | AK057536.1 | - | 1 | 105 | ||
AK124283.1 | - | 1 | 1163 | |||
AY605659.1 | - | 1 | 421 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
