siRNA Details
Gene Symbol: PKHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138694.3 | NP_619639.3 | 29 | 3603 | ||
NM_170724.2 | NP_733842.2 | 29 | 3603 | |||
XM_011514680.2 | XP_011512982.1 | 29 | 3947 | |||
XM_011514682.2 | XP_011512984.1 | 29 | 3569 | |||
XM_011514683.2 | XP_011512985.1 | 29 | 3569 | |||
XM_011514684.2 | XP_011512986.1 | 21 | 2713 | |||
XM_011514685.1 | XP_011512987.1 | 29 | 3603 | |||
XM_011514686.2 | XP_011512988.1 | 29 | 3603 | |||
XM_011514687.1 | XP_011512989.1 | 29 | 3603 | |||
XM_011514688.2 | XP_011512990.1 | 29 | 3569 | |||
XM_017010944.1 | XP_016866433.1 | 29 | 3429 | |||
XM_017010945.1 | XP_016866434.1 | 28 | 3494 | |||
XM_017010946.1 | XP_016866435.1 | 29 | 3569 | |||
XM_017010947.1 | XP_016866436.1 | 29 | 3569 | |||
XM_017010948.1 | XP_016866437.1 | 20 | 3387 | |||
XM_017010949.1 | XP_016866438.1 | 11 | 2566 | |||
XM_017010950.1 | XP_016866439.1 | 29 | 3603 | |||
XM_017010951.1 | XP_016866440.1 | 29 | 3569 | |||
XM_017010952.1 | XP_016866441.1 | 29 | 3569 | |||
GenBank mRNA | AF480064.1 | - | 29 | 3549 | ||
AY074797.1 | - | 29 | 3603 | |||
AY092083.1 | - | 29 | 3589 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
