siRNA Details
Gene Symbol: BLOC1S3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212550.4 | NP_997715.1 | 2 | 167 | ||
GenBank mRNA | AK296695.1 | - | 2 | 165 | ||
AY531266.1 | - | 1 | 71 | |||
BC151151.1 | - | 1 | 145 | 9021441 | ||
BC151152.1 | - | 1 | 143 | 9021442 | ||
DC422050.1 | - | 2 | 167 | |||
DN996082.1 | - | 2 | 147 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
