siRNA Details
Gene Symbol: FERMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134999.1 | NP_001128471.1 | 11 | 1493 | ||
NM_001135000.1 | NP_001128472.1 | 11 | 1493 | |||
NM_006832.2 | NP_006823.1 | 11 | 1493 | |||
XM_005267285.2 | XP_005267342.1 | 12 | 1621 | |||
XM_006720008.2 | XP_006720071.1 | 12 | 1621 | |||
XM_006720009.2 | XP_006720072.1 | 12 | 1621 | |||
XM_006720010.2 | XP_006720073.1 | 11 | 1588 | |||
GenBank mRNA | AF443279.1 | - | 11 | 1388 | ||
AK291738.1 | - | 11 | 1446 | |||
BC017327.2 | - | 11 | 1493 | 4547604 | ||
BX161467.1 | - | 11 | 1414 | |||
EU979385.1 | - | 10 | 1307 | |||
Z24725.1 | - | 11 | 1429 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
