siRNA Details
Gene Symbol: FERMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134999.1 | NP_001128471.1 | 3 | 357 | ||
NM_001135000.1 | NP_001128472.1 | 3 | 357 | |||
NM_006832.2 | NP_006823.1 | 3 | 357 | |||
XM_005267285.2 | XP_005267342.1 | 3 | 452 | |||
XM_006720008.2 | XP_006720071.1 | 3 | 452 | |||
XM_006720009.2 | XP_006720072.1 | 3 | 452 | |||
XM_006720010.2 | XP_006720073.1 | 3 | 452 | |||
GenBank mRNA | AF443279.1 | - | 3 | 252 | ||
AK291738.1 | - | 3 | 310 | |||
BC017327.2 | - | 3 | 357 | 4547604 | ||
BX161467.1 | - | 3 | 278 | |||
EU979385.1 | - | 2 | 171 | |||
Z24725.1 | - | 3 | 293 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
