siRNA Details
Gene Symbol: COL18A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030582.3 | NP_085059.2 | 37 | 3782 | ||
NM_130444.2 | NP_569711.2 | 37 | 4487 | |||
NM_130445.3 | NP_569712.2 | 38 | 3315 | |||
GenBank mRNA | AF018081.1 | - | 37 | 3801 | ||
AF018082.1 | - | 38 | 3280 | |||
AK098216.1 | - | 4 | 1405 | |||
AK130835.1 | - | 5 | 432 | |||
BC033715.1 | - | 26 | 1663 | |||
BC063833.1 | - | 38 | 3256 | 6181818 | ||
L22548.1 | - | 19 | 1265 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
